Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103352.1 | NP_001096822.1 | 4, 5 | 951 | ||
XM_006255086.2 | XP_006255148.1 | 8, 9 | 1253 | |||
XM_006255087.2 | XP_006255149.1 | 8, 9 | 1253 | |||
XM_006255088.2 | XP_006255150.1 | 8, 9 | 1253 | |||
XM_008772286.1 | XP_008770508.1 | 8, 9 | 1250 | |||
XM_008772287.1 | XP_008770509.1 | 6, 7 | 932 | |||
XM_008772288.1 | XP_008770510.1 | 8, 9 | 1243 | |||
XM_008772289.1 | XP_008770511.1 | 7, 8 | 1026 | |||
XM_008772290.1 | XP_008770512.1 | 6, 7 | 1115 | |||
XM_008772291.1 | XP_008770513.1 | 7, 8 | 1093 | |||
XM_008772292.1 | XP_008770514.1 | 5, 6 | 722 | |||
XM_008772293.1 | XP_008770515.1 | 4, 5 | 729 | |||
XM_008772294.1 | XP_008770516.1 | 5, 6 | 2888 | |||
XM_008772295.1 | XP_008770517.1 | 6, 7 | 921 | |||
XM_008772296.1 | XP_008770518.1 | 4, 5 | 2744 | |||
GenBank mRNA | BC128748.1 | - | 4, 5 | 951 | 7099524 |