Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134708.1 | NP_001128180.1 | 2, 3 | 338 | ||
XM_006233439.2 | XP_006233501.1 | 3, 4 | 778 | |||
XM_006233440.2 | XP_006233502.1 | 3, 4 | 922 | |||
XM_006233441.2 | XP_006233503.1 | 3, 4 | 906 | |||
XM_006233442.2 | XP_006233504.1 | 2, 3 | 6766 | |||
XM_006233443.2 | XP_006233505.1 | 3, 4 | 910 | |||
XM_006233444.2 | XP_006233506.1 | 1, 2 | 6686 | |||
XM_006233445.2 | XP_006233507.1 | 2, 3 | 830 | |||
XM_006233446.2 | XP_006233508.1 | 2, 3 | 6762 | |||
XM_006233447.2 | XP_006233509.1 | 2, 3 | 6760 | |||
XM_006233448.2 | XP_006233510.1 | 1, 2 | 6685 | |||
XM_006233449.2 | XP_006233511.1 | 1, 2 | 6685 | |||
XM_008761524.1 | XP_008759746.1 | 2, 3 | 6762 | |||
XM_008761525.1 | XP_008759747.1 | 3, 4 | 1050 | |||
GenBank mRNA | BC166823.1 | - | 2, 3 | 338 | 8370140 |