Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irgc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014016.1 | NP_001014038.1 | 2 | 607 | ||
XM_006228425.2 | XP_006228487.1 | 2 | 656 | |||
XM_006228426.2 | XP_006228488.1 | 2 | 536 | |||
XM_006228427.2 | XP_006228489.1 | 2 | 451 | |||
XM_008758927.1 | XP_008757149.1 | 2 | 520 | |||
GenBank mRNA | BC079034.1 | - | 2 | 554 | 7114307 | |
BC079061.1 | - | 2 | 607 | 7115757 |