Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730284.4 | XP_002730330.1 | 7 | 2051 | ||
XM_006257246.2 | XP_006257308.1 | 6 | 1800 | |||
XM_006257247.1 | XP_006257309.1 | 8 | 1126 | |||
XM_006257248.1 | XP_006257310.1 | 9 | 1207 | |||
XM_006257250.2 | XP_006257312.1 | 8 | 1081 | |||
XM_008773457.1 | XP_008771679.1 | 7 | 4460 | |||
XM_008773458.1 | XP_008771680.1 | 7 | 3192 |