Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lins | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229357.2 | XP_006229419.1 | 8 | 2089 | ||
XM_006229358.2 | XP_006229420.1 | 7 | 1883 | |||
XM_008759572.1 | XP_008757794.1 | 8 | 2211 | |||
XM_008759573.1 | XP_008757795.1 | 8 | 2170 | |||
XM_008759574.1 | XP_008757796.1 | 7 | 2266 |