Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rela | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199267.2 | NP_954888.1 | 11 | 1671 | ||
GenBank mRNA | AY307375.1 | - | 10 | 1610 | ||
BC079457.1 | - | 11 | 1671 | 7121610 |