Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199103.1 | NP_954534.1 | 6 | 807 | ||
XM_006229580.2 | XP_006229642.1 | 8 | 2055 | |||
XM_006229584.2 | XP_006229646.1 | 8 | 1041 | |||
XM_006229585.2 | XP_006229647.1 | 7 | 1001 | |||
XM_008773600.1 | XP_008771822.1 | 7 | 5115 | |||
XM_008773601.1 | XP_008771823.1 | 8 | 2058 | |||
XM_008773602.1 | XP_008771824.1 | 8 | 2072 | |||
XM_008773603.1 | XP_008771825.1 | 8 | 2063 | |||
XM_008773604.1 | XP_008771826.1 | 7 | 5112 | |||
XM_008773605.1 | XP_008771827.1 | 7 | 5112 | |||
XM_008773606.1 | XP_008771828.1 | 7 | 5109 | |||
XM_008773607.1 | XP_008771829.1 | 8 | 5155 | |||
XM_008773608.1 | XP_008771830.1 | 8 | 1045 | |||
XM_008773609.1 | XP_008771831.1 | 8 | 1253 | |||
XM_008773610.1 | XP_008771832.1 | 7 | 1005 | |||
GenBank mRNA | AY392762.1 | - | 6 | 807 |