Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199103.1 | NP_954534.1 | 1 | 115 | ||
XM_006229580.2 | XP_006229642.1 | 3 | 1363 | |||
XM_006229584.2 | XP_006229646.1 | 3 | 349 | |||
XM_006229585.2 | XP_006229647.1 | 2 | 309 | |||
XM_008773600.1 | XP_008771822.1 | 2 | 4420 | |||
XM_008773601.1 | XP_008771823.1 | 3 | 1363 | |||
XM_008773602.1 | XP_008771824.1 | 3 | 1377 | |||
XM_008773603.1 | XP_008771825.1 | 3 | 1368 | |||
XM_008773604.1 | XP_008771826.1 | 2 | 4420 | |||
XM_008773605.1 | XP_008771827.1 | 2 | 4420 | |||
XM_008773606.1 | XP_008771828.1 | 2 | 4420 | |||
XM_008773607.1 | XP_008771829.1 | 3 | 4460 | |||
XM_008773608.1 | XP_008771830.1 | 3 | 350 | |||
XM_008773609.1 | XP_008771831.1 | 3 | 558 | |||
XM_008773610.1 | XP_008771832.1 | 2 | 310 | |||
GenBank mRNA | AY392762.1 | - | 1 | 115 |