Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231255.2 | XP_006231317.1 | 16 | 1956 | ||
XM_006231256.2 | XP_006231318.1 | 16 | 1954 | |||
XM_219778.8 | XP_219778.5 | 16 | 1956 |