Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107595.1 | NP_001101065.1 | 9 | 1143 |
Gene Symbol: LOC100911700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760501.1 | XP_008758723.1 | 7 | 798 |