Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107599.1 | NP_001101069.1 | 5 | 2599 | ||
XM_008760429.1 | XP_008758651.1 | 5 | 2659 | |||
XM_008760430.1 | XP_008758652.1 | 5 | 659 | |||
XM_008760431.1 | XP_008758653.1 | 5 | 777 |