Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107609.2 | NP_001101079.2 | 30 | 4627 | ||
XM_006231281.2 | XP_006231343.1 | 27 | 4703 | |||
XM_006231283.2 | XP_006231345.1 | 27 | 4685 | |||
XM_006231284.2 | XP_006231346.1 | 27 | 4583 | |||
XM_006231285.2 | XP_006231347.1 | 27 | 4565 | |||
XM_006231286.1 | XP_006231348.1 | 23 | 4208 | |||
XM_008760328.1 | XP_008758550.1 | 27 | 4860 | |||
XM_008760329.1 | XP_008758551.1 | 27 | 4692 |