Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107613.3 | NP_001101083.1 | 17 | 2686 | ||
XM_006256193.2 | XP_006256255.1 | 18 | 2761 | |||
XM_006256195.2 | XP_006256257.1 | 17 | 2698 | |||
XM_006256196.2 | XP_006256258.1 | 18 | 2761 | |||
XM_006256197.2 | XP_006256259.1 | 17 | 2698 | |||
XM_008772813.1 | XP_008771035.1 | 18 | 2761 | |||
XM_008772814.1 | XP_008771036.1 | 10 | 2622 | |||
XM_008772815.1 | XP_008771037.1 | 8 | 1199 |