Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mypn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107628.1 | NP_001101098.1 | 17 | 3635 | ||
XM_006256374.2 | XP_006256436.1 | 19 | 3944 |