Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107634.1 | NP_001101104.1 | 3, 4 | 584 | ||
XM_008772985.1 | XP_008771207.1 | 2, 3 | 199 |