Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mctp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760610.1 | XP_008758832.1 | 9 | 1509 | ||
XM_008760611.1 | XP_008758833.1 | 8 | 1470 | |||
XM_008760612.1 | XP_008758834.1 | 10 | 4189 | |||
XM_008760613.1 | XP_008758835.1 | 9 | 4053 | |||
XM_008760614.1 | XP_008758836.1 | 9 | 1505 | |||
XM_008760615.1 | XP_008758837.1 | 8 | 1466 | |||
XM_008760616.1 | XP_008758838.1 | 5 | 3659 | |||
XM_008760617.1 | XP_008758839.1 | 5 | 540 | |||
GenBank mRNA | FQ232558.1 | - | 8 | 1089 |