Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107651.1 | NP_001101121.1 | 4 | 622 | ||
XM_008760729.1 | XP_008758951.1 | 4 | 562 |