Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035251.1 | NP_001030328.1 | 6 | 670 | ||
XM_006231929.2 | XP_006231991.1 | 7 | 797 | |||
XM_006231930.2 | XP_006231992.1 | 8 | 861 | |||
XM_006231931.2 | XP_006231993.1 | 7 | 838 | |||
XM_006231932.2 | XP_006231994.1 | 7 | 928 | |||
XM_006231933.2 | XP_006231995.1 | 7 | 887 | |||
XM_006231934.2 | XP_006231996.1 | 6 | 2872 | |||
XM_006231936.2 | XP_006231998.1 | 6 | 2857 | |||
XM_006231937.2 | XP_006231999.1 | 6 | 638 | |||
XM_006231939.2 | XP_006232001.1 | 6 | 550 | |||
GenBank mRNA | BC092662.1 | - | 5 | 532 | ||
BC105869.1 | - | 6 | 670 | 7442690 | ||
JQ013729.1 | - | 6 | 721 |