Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rictor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760806.1 | XP_008759028.1 | 16, 17 | 2803 | ||
XM_008760807.1 | XP_008759029.1 | 19, 20 | 1713 | |||
XM_008760808.1 | XP_008759030.1 | 18, 19 | 1676 |