Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drosha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107655.2 | NP_001101125.2 | 24 | 3228 | ||
XM_006232049.2 | XP_006232111.1 | 21 | 3051 | |||
XM_006232050.2 | XP_006232112.1 | 21 | 3027 | |||
GenBank mRNA | FQ212690.1 | - | 3 | 162 | ||
FQ215440.1 | - | 4 | 222 |