Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107666.2 | NP_001101136.2 | 3, 4 | 546 | ||
XM_006232257.2 | XP_006232319.1 | 3, 4 | 596 |