Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_227201.8 | XP_227201.4 | 14 | 3135 | ||
GenBank mRNA | FQ218213.1 | - | 13 | 1780 |