Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135717.1 | NP_001129189.1 | 4 | 883 | ||
XM_006232530.2 | XP_006232592.1 | 4 | 878 |