Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012079.1 | NP_001012079.1 | 6, 7 | 654 | ||
XM_006232643.2 | XP_006232705.1 | 11, 12 | 1432 | |||
XM_006232644.2 | XP_006232706.1 | 12, 13 | 1549 | |||
XM_006232646.2 | XP_006232708.1 | 10, 11 | 1009 | |||
XM_006232647.2 | XP_006232709.1 | 9, 10 | 853 | |||
XM_006232648.2 | XP_006232710.1 | 9, 10 | 804 | |||
XM_006232651.2 | XP_006232713.1 | 6, 7 | 942 | |||
XM_006232652.2 | XP_006232714.1 | 6, 7 | 761 | |||
XM_006232653.2 | XP_006232715.1 | 6, 7 | 649 | |||
XM_006232654.2 | XP_006232716.1 | 6, 7 | 1458 | |||
XM_008761151.1 | XP_008759373.1 | 12, 13 | 1544 | |||
XM_008761152.1 | XP_008759374.1 | 7, 8 | 1532 | |||
GenBank mRNA | BC090078.1 | - | 6, 7 | 654 | 7125049 | |
FQ212011.1 | - | 7, 8 | 627 |