Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134504.1 | NP_001127976.1 | 2 | 1130 | ||
XM_006234313.2 | XP_006234375.1 | 2 | 1096 | |||
XM_006234314.2 | XP_006234376.1 | 2 | 1592 | |||
XM_006234316.2 | XP_006234378.1 | 2 | 1337 |