Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107734.1 | NP_001101204.1 | 2086 | |||
XM_008761896.1 | XP_008760118.1 | 20 | 2307 | |||
XM_008761897.1 | XP_008760119.1 | 3539 | ||||
XM_008761898.1 | XP_008760120.1 | 20 | 2468 | |||
XM_008761899.1 | XP_008760121.1 | 2089 | ||||
GenBank mRNA | FQ229141.1 | - | 180 |
Gene Symbol: LOC103690017 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234342.2 | XP_006234404.2 | 20 | 2089 | ||
XM_008761952.1 | XP_008760174.1 | 20 | 3539 |