Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp385b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107736.1 | NP_001101206.1 | 6 | 2057 | ||
XM_006234414.2 | XP_006234476.1 | 7 | 1716 | |||
XM_006234416.2 | XP_006234478.1 | 6 | 1677 | |||
XM_006234419.2 | XP_006234481.1 | 7 | 2873 | |||
XM_006234420.2 | XP_006234482.1 | 7 | 1242 |