Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107742.1 | NP_001101212.1 | 7 | 1060 | ||
GenBank mRNA | BC161926.1 | - | 7 | 1021 | 8373798 |