Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107747.2 | NP_001101217.2 | 7 | 720 | ||
XM_006234551.1 | XP_006234613.1 | 8 | 757 | |||
XM_006234552.2 | XP_006234614.1 | 8 | 787 | |||
XM_006234553.2 | XP_006234615.1 | 8 | 757 | |||
XM_006234554.1 | XP_006234616.1 | 9 | 799 | |||
XM_006234556.1 | XP_006234618.1 | 8 | 734 | |||
XM_006234557.2 | XP_006234619.1 | 8 | 764 | |||
XM_006234558.2 | XP_006234620.1 | 7 | 705 | |||
XM_006234559.2 | XP_006234621.1 | 5 | 2143 | |||
XM_006234560.2 | XP_006234622.1 | 7 | 746 | |||
GenBank mRNA | EV768858.1 | - | 3 | 191 | ||
FQ224002.1 | - | 7 | 680 |