Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107747.2 | NP_001101217.2 | 13 | 1285 | ||
XM_006234551.1 | XP_006234613.1 | 14 | 1322 | |||
XM_006234552.2 | XP_006234614.1 | 14 | 1352 | |||
XM_006234553.2 | XP_006234615.1 | 14 | 1322 | |||
XM_006234554.1 | XP_006234616.1 | 15 | 1364 | |||
XM_006234556.1 | XP_006234618.1 | 14 | 1299 | |||
XM_006234557.2 | XP_006234619.1 | 14 | 1329 | |||
XM_006234558.2 | XP_006234620.1 | 13 | 1270 | |||
XM_006234559.2 | XP_006234621.1 | 11 | 2708 | |||
XM_006234560.2 | XP_006234622.1 | 13 | 1311 | |||
GenBank mRNA | DN932805.1 | - | 5 | 445 | ||
FQ224002.1 | - | 13 | 1245 |