Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ganc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145840.1 | NP_001139312.1 | 7, 8 | 1752 | ||
XM_006234721.2 | XP_006234783.1 | 7, 8 | 639 | |||
XM_006234722.2 | XP_006234784.1 | 7, 8 | 639 |