Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppip5k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080783.1 | NP_001074252.1 | 9 | 1113 | ||
XM_006234838.2 | XP_006234900.1 | 10 | 1187 | |||
XM_006234840.2 | XP_006234902.1 | 10 | 1307 | |||
XM_006234841.2 | XP_006234903.1 | 10 | 1582 | |||
XM_006234843.2 | XP_006234905.1 | 10 | 1187 | |||
XM_006234844.2 | XP_006234906.1 | 10 | 1187 | |||
XM_006234845.2 | XP_006234907.1 | 10 | 1187 | |||
XM_006234848.2 | XP_006234910.1 | 10 | 1187 | |||
XM_006234849.2 | XP_006234911.1 | 10 | 1187 | |||
XM_006234850.2 | XP_006234912.1 | 10 | 1187 | |||
XM_006234851.2 | XP_006234913.1 | 10 | 1187 | |||
XM_006234852.2 | XP_006234914.1 | 10 | 1187 | |||
XM_008762170.1 | XP_008760392.1 | 3 | 366 |