Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serinc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110811.1 | NP_001104281.1 | 6 | 965 | ||
XM_006234853.2 | XP_006234915.1 | 6 | 1047 | |||
GenBank mRNA | EU220432.1 | - | 2 | 43 |