Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEWGENE_1311061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234999.2 | XP_006235061.1 | 4 | 2487 | ||
XM_006235000.2 | XP_006235062.1 | 4 | 2493 | |||
XM_008762233.1 | XP_008760455.1 | 4 | 2492 | |||
XM_008762235.1 | XP_008760457.1 | 4 | 760 |
Gene Symbol: Zc3h6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107772.1 | NP_001101242.1 | 924 |