Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107795.1 | NP_001101265.1 | 5 | 566 | ||
XM_006235404.2 | XP_006235466.1 | 6 | 642 | |||
XM_008762342.1 | XP_008760564.1 | 6 | 653 | |||
GenBank mRNA | FQ228046.1 | - | 2 | 98 | ||
FQ232914.1 | - | 2 | 98 |