Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173511.1 | NP_001166982.1 | 26 | 2639 | ||
XM_006236037.2 | XP_006236099.1 | 26 | 2670 | |||
XM_008762704.1 | XP_008760926.1 | 26 | 2673 | |||
GenBank mRNA | BC091288.1 | - | 11 | 1234 |
Gene Symbol: LOC100911994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749697.3 | XP_003749745.2 | 6 | 1897 |