Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107862.1 | NP_001101332.1 | 1 | 142 | ||
XM_006236552.2 | XP_006236614.1 | 1 | 239 | |||
GenBank mRNA | BC166444.1 | - | 1 | 164 | 8363316 |