Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107868.1 | NP_001101338.1 | 22 | 4132 | ||
XM_006236749.2 | XP_006236811.1 | 22 | 4275 | |||
XM_006236750.2 | XP_006236812.1 | 21 | 4206 | |||
XM_006236751.2 | XP_006236813.1 | 22 | 4275 | |||
XM_006236753.2 | XP_006236815.1 | 20 | 4122 | |||
XM_006236754.2 | XP_006236816.1 | 22 | 4294 |