Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170584.1 | NP_001164055.1 | 5 | 598 | ||
XM_006237384.2 | XP_006237446.1 | 5 | 599 | |||
XM_006237385.2 | XP_006237447.1 | 5 | 529 | |||
GenBank mRNA | CB809540.1 | - | 1 | 44 | ||
CK477113.1 | - | 5 | 467 |
Gene Symbol: LOC103690024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763277.1 | XP_008761499.1 | 530 |