Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135780.1 | NP_001129252.1 | 12 | 1715 | ||
XM_006237954.2 | XP_006238016.1 | 12 | 1767 |