Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100910771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750753.3 | XP_003750801.1 | 6 | 688 | ||
XM_006245614.2 | XP_006245676.1 | 6 | 688 |
Gene Symbol: Map3k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107920.2 | NP_001101390.2 | 6 | 712 | ||
XM_006237966.2 | XP_006238028.1 | 6 | 1904 |