Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014063.1 | NP_001014085.1 | 3 | 1097 | ||
GenBank mRNA | BC089827.1 | - | 3 | 1097 | 7321838 |