Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270984.1 | NP_001257913.1 | 14, 15 | 1711 | ||
GenBank mRNA | FQ212419.1 | - | 5, 6 | 2432 | ||
FQ222909.1 | - | 11, 12 | 1313 |