Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270984.1 | NP_001257913.1 | 12 | 1435 | ||
GenBank mRNA | FQ212419.1 | - | 4 | 1748 | ||
FQ222909.1 | - | 9 | 1037 |