Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107956.1 | NP_001101426.1 | 8 | 1088 | ||
XM_008763676.1 | XP_008761898.1 | 8 | 1744 | |||
GenBank mRNA | EF187254.1 | - | 8 | 1090 |