Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107958.2 | NP_001101428.2 | 5 | 1045 | ||
XM_006238615.2 | XP_006238677.1 | 5 | 911 | |||
XM_006238617.1 | XP_006238679.1 | 5 | 1119 | |||
XM_006238618.2 | XP_006238680.1 | 4 | 763 | |||
XM_008763967.1 | XP_008762189.1 | 5 | 1430 |