Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HEYL SIRNA Assays ›
Gene Symbol:
Heyl
Gene Name:
hes-related family bHLH transcription factor with YRPW motif-like
Gene Aliases:
-
Chromosome Location:
Chr.5: 140923914 - 140940905 on Build Rnor_6.0
UniGene ID:
Rn.22252
Species:
Rat
siRNA ID
RSS320339
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Heyl
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001107977.1
NP_001101447.1
5
1265
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
outflow tract morphogenesis
atrioventricular valve morphogenesis
pulmonary valve morphogenesis
epithelial to mesenchymal transition involved in endocardial cushion formation
endocardial cushion morphogenesis
cardiac ventricle morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
Notch signaling pathway
negative regulation of gene expression
mesenchymal cell development
skeletal muscle cell differentiation
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cardiac epithelial to mesenchymal transition
ventricular septum morphogenesis
negative regulation of androgen receptor signaling pathway
negative regulation of androgen receptor activity
RNA polymerase II core promoter sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter sequence-specific
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription corepressor activity
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
microsatellite binding
protein homodimerization activity
sequence-specific DNA binding
protein heterodimerization activity
protein dimerization activity
AF-1 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0