Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108004.1 | NP_001101474.1 | 2 | 255 | ||
XM_006238070.2 | XP_006238132.1 | 2 | 288 |