Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108010.2 | NP_001101480.1 | 3 | 982 | ||
GenBank mRNA | BC168944.1 | - | 3 | 982 | 8371512 |