Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170347.1 | NP_001163818.1 | 23, 24 | 4469 | ||
XM_006240299.2 | XP_006240361.1 | 23, 24 | 4480 | |||
XM_006240300.2 | XP_006240362.1 | 23, 24 | 4477 | |||
XM_006240301.1 | XP_006240363.1 | 24, 25 | 4516 | |||
XM_006240302.2 | XP_006240364.1 | 21, 22 | 4162 | |||
XM_008764788.1 | XP_008763010.1 | 22, 23 | 4390 | |||
XM_008764789.1 | XP_008763011.1 | 23, 24 | 5876 |